Learn More
VWR International Freestanding Micro Centrifuge Tubes, Non-sterile, 2 mL, 500/pk
Catalog No. NC1982756
Encompass
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents. They feature universal screw threads for use with most popular brand screw caps. Tubes only.
Catalog No. NC1982756 Supplier VWR International Supplier No. 16466032
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents. They feature universal screw threads for use with most popular brand screw caps. Tubes only.