Learn More
TWD Scientific LLC Tubes, Micro-Centrifuge, Twdbiox, 2.0Ml, Polypropylene, Non-Sterile, Positive Cap Closure, Marked Graduations, Frosted Writing Are, 5000/Case.
SDP
Catalog No. 502285010 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 2.0Ml Clear Polypropylene Non-Sterile (Rnase, Dnase, Dna, Pyrogen Free) Microcentrifuge Tubes With Positive Cap Closure. Tubes Are Boil Proof And Rated For Rcf Up To 20,000Xg. Tubes Come With Marked Graduations And Frosted Writing Area. 500 Tubes/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5010 Supplier TWD Scientific LLC Supplier No. BX801310
BioX 2.0Ml Clear Polypropylene Non-Sterile (Rnase, Dnase, Dna, Pyrogen Free) Microcentrifuge Tubes With Positive Cap Closure. Tubes Are Boil Proof And Rated For Rcf Up To 20,000Xg. Tubes Come With Marked Graduations And Frosted Writing Area. 500 Tubes/Pack, 10 Packs/Case, Price/Case