Learn More
Electron Microscopy Sciences Micro Cleaning Solution 1 QT
SDP
Catalog No. 501899445
Encompass_Preferred
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber. You can work with any concentration. Contains no phosphates and it is biodegradable. Even removes radioactive contaminants. Not to be used on zinc, magnesium, or aluminum.
Catalog No. 50-189-9445 Supplier Electron Microscopy Sciences Supplier No. 71055
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber. You can work with any concentration. Contains no phosphates and it is biodegradable. Even removes radioactive contaminants. Not to be used on zinc, magnesium, or aluminum.