Learn More
Electron Microscopy Sciences Micro Cleaning Solution 1 GL
SDP
Catalog No. 501899446
Encompass_Preferred
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber. You can work with any concentration. Contains no phosphates and it is biodegradable. Even removes radioactive contaminants. Not to be used on zinc, magnesium, or aluminum.
Catalog No. 50-189-9446 Supplier Electron Microscopy Sciences Supplier No. 71056
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber. You can work with any concentration. Contains no phosphates and it is biodegradable. Even removes radioactive contaminants. Not to be used on zinc, magnesium, or aluminum.