Learn More
Andwin Scientific MICRO CLEANING SOLN 12X1L
SDP
Catalog No. NC0114649
Encompass_Preferred
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged in a 1 liter plastic bottle and is sold in a case of 12.
This item is not returnable. View return policy
Catalog No. NC0114649 Supplier Andwin Scientific Supplier No. 89177068
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged in a 1 liter plastic bottle and is sold in a case of 12.