Learn More
Andwin Scientific MICRO CLEANING SOLUTION 1QT
SDP
Catalog No. 50930737
Encompass_Preferred
The Micro Cleaning Solution is made to sanitize glass, most metals, plastics, and rubber in order to remove pollutants and even radioactive contaminants. This product does not have phosphates. This solution should not be applied on zinc, magnesium, or aluminum. It is sold in a quantity of 1 quart.
Catalog No. 50-930-737 Supplier Andwin Scientific Supplier No. 71054
The Micro Cleaning Solution is made to sanitize glass, most metals, plastics, and rubber in order to remove pollutants and even radioactive contaminants. This product does not have phosphates. This solution should not be applied on zinc, magnesium, or aluminum. It is sold in a quantity of 1 quart.