Learn More
Roboz Surgical MICRO CLIP 11MM ANG
SDP
Catalog No. 5000574593
Encompass
This micro vascular clip is a surgical instrument designed specifically for animal research applications. It features an 11 mm length with a 55-degree angle to facilitate precise tissue manipulation and vessel occlusion in delicate experimental procedures.
  • Designed for animal research use only
  • Equipped with 1.5 x 5 mm angled jaws
  • Constructed for precision handling
Catalog No. 50-005-74593 Supplier Roboz Surgical Supplier No. RS6473
This micro vascular clip is a surgical instrument designed specifically for animal research applications. It features an 11 mm length with a 55-degree angle to facilitate precise tissue manipulation and vessel occlusion in delicate experimental procedures.
  • Designed for animal research use only
  • Equipped with 1.5 x 5 mm angled jaws
  • Constructed for precision handling