Learn More
Stoelting™ Micro-Clip Holding Forceps
SDP
Catalog No. 10001367 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-001-367 Neuroscience
1 options
Catalog No. 10-001-367 Supplier Stoelting™ Supplier No. 5212006
 Restricted Item
Add to Cart
Add to Cart
Stoelting™ Micro-Clip Holding Forceps are micro tweezers that are useful in a variety of different applications including micro dissection of tissue and vessels, and processing tissue in electron microscopy procedures.
Specifications
For Use With (Application) Neuroscience
Length (English) 4.75 in.