Learn More
Roboz Surgical MICRO CLIP STR 1X8MM
SDP
Catalog No. NC9432791
Encompass
Micro Clip; Straight; 70 Grams Pressure; 1mm Clip Width; 8mm Jaw Length. The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and it
Catalog No. NC9432791 Supplier Roboz Surgical Supplier No. RS5424
Micro Clip; Straight; 70 Grams Pressure; 1mm Clip Width; 8mm Jaw Length. The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and it