Learn More
Roboz Surgical MICRO CLIP STR .8X6MM
SDP
Catalog No. NC9432790
Encompass
Micro Clip; Straight; 70 Grams Pressure; 0.75mm Clip Width; 6mm Jaw Length The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and i
Catalog No. NC9432790 Supplier Roboz Surgical Supplier No. RS5422
Micro Clip; Straight; 70 Grams Pressure; 0.75mm Clip Width; 6mm Jaw Length The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and i