Learn More
Electron Microscopy Sciences Micro Centrifuge Tube 96 Place Rack
SDP
Catalog No. 501899866
Encompass_Preferred
This sturdy polypropylene rack holds 96 tubes. The reversible rack holds 1.5/2.0 ml tubes on one side and 0.5 ml tubes on the other in an 8 x 12 format. Easy access to each tube even when full. The transparent polypropylene lid has a snug fit and is tall enough to store most screw-cap microcentrifuge tubes. Withstands freezing (-90 degree C) and autoclaving. Frosted labeling area on four sides for easy sample identificatio
Catalog No. 50-189-9866 Supplier Electron Microscopy Sciences Supplier No. 7237506
This sturdy polypropylene rack holds 96 tubes. The reversible rack holds 1.5/2.0 ml tubes on one side and 0.5 ml tubes on the other in an 8 x 12 format. Easy access to each tube even when full. The transparent polypropylene lid has a snug fit and is tall enough to store most screw-cap microcentrifuge tubes. Withstands freezing (-90 degree C) and autoclaving. Frosted labeling area on four sides for easy sample identificatio