Learn More
Analytical Sales & Services Micro Column Connection Kit
Catalog No. 5000806132
Encompass
Micro Column Connection Kit - Kit includes:\n\n• PEEK one-piece fittings\n• Tubing sleeves\n• Fused silica capillary tubing, 360μm OD
Catalog No. 50-008-06132 Supplier Analytical Sales & Services Supplier No. ADVMFKIT
Micro Column Connection Kit - Kit includes:\n\n• PEEK one-piece fittings\n• Tubing sleeves\n• Fused silica capillary tubing, 360μm OD