Learn More
Metrohm USA MICRO COMBINATION ELECTRODE-16
SDP
Catalog No. NC0344231 Shop All Metrohm USA Products
Encompass
Routine use on sample changers and in small measuring vessels for acid-base titrations.
Catalog No. NC0344231 Supplier Metrohm USA Supplier No. 60234110
Routine use on sample changers and in small measuring vessels for acid-base titrations.