Learn More
Ted Pella Inc Micro Cover Glasses, 22 x 22mm x 0.13 - 0.16mm thick, 1 oz/pk
SDP
Catalog No. NC9448018
Encompass
Individually selected, superior quality, cleaned, cut, sorted and made of especially processed borosilicate glass, and not sterile.
Catalog No. NC9448018 Supplier Ted Pella Inc Supplier No. 260140
Individually selected, superior quality, cleaned, cut, sorted and made of especially processed borosilicate glass, and not sterile.