Learn More
Ted Pella Inc Micro Cover Glasses, 22 x 22mm x 0.16 - 0.19mm thick, 1 oz/pk
SDP
Supplier:  Ted Pella Inc 260148
Encompass
Individually selected, superior quality, cleaned, cut, sorted and made of especially processed borosilicate glass, and not sterile.
Catalog No. NC1466408