Learn More
Andwin Scientific VWR Micro Cover Glasses Square, 1oz
SDP
Catalog No. NC1397033
Encompass_Preferred
VWR Micro Cover Glasses Square No. 1-1/2 No. 1-1/2 thickness is 0.16 to 0.19mm. 22x22mm. These cover glasses offer maximum clarity superior corrosion resistance and uniform thickness and size. They are free of striae and oily film.
Catalog No. NC1397033 Supplier Andwin Scientific Supplier No. 48366227
VWR Micro Cover Glasses Square No. 1-1/2 No. 1-1/2 thickness is 0.16 to 0.19mm. 22x22mm. These cover glasses offer maximum clarity superior corrosion resistance and uniform thickness and size. They are free of striae and oily film.