Learn More
Andwin Scientific MICRO COVERGLASS 22X22 1 OZ
SDP
Supplier:  Andwin Scientific 7221010
Encompass_Preferred
The micro cover glasses are made from clear white, borosilicate glass that provide superior clarity. This product has a size of 22 x 22 mm and thickness of 0.17-0.25 mm. It also features resistance against corrosion and a perfectly flat design. This is sold in a 1 ounce pack of 156 cover glasses.
Catalog No. 50-948-956