Learn More
Andwin Scientific MICRO COVERGLASS 22X22 10 OZ
SDP
Catalog No. 50949035
Encompass_Preferred
MICRO COVERGLASS 22X22 10 OZ. Thickness #1-1/2 (.16-.19 mm)
Catalog No. 50-949-035 Supplier Andwin Scientific Supplier No. 7220410
MICRO COVERGLASS 22X22 10 OZ. Thickness #1-1/2 (.16-.19 mm)