Learn More
Andwin Scientific MICRO COVERGLASS 22X22 10 OZ
SDP
Catalog No. 50949031
Encompass_Preferred
MICRO COVERGLASS 22X22 10 OZ. Thickness #1 (.13-.17 mm)
Catalog No. 50-949-031 Supplier Andwin Scientific Supplier No. 7220011
MICRO COVERGLASS 22X22 10 OZ. Thickness #1 (.13-.17 mm)