Learn More
Andwin Scientific MICRO COVERGLASS 22X30 1 OZ
SDP
Catalog No. 50948953
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness. MICRO COVERGLASS 22X30 1 OZ, #1 (.16-.19 mm)
Catalog No. 50-948-953 Supplier Andwin Scientific Supplier No. 7220402
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness. MICRO COVERGLASS 22X30 1 OZ, #1 (.16-.19 mm)