Learn More
Andwin Scientific Micro Cover Glasses 22x30 mm 10OZ
SDP
Catalog No. 50949036
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness. #1½ (.16-.19 mm) 22x30 mm , 10oz
Catalog No. 50-949-036 Supplier Andwin Scientific Supplier No. 7220420
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness. #1½ (.16-.19 mm) 22x30 mm , 10oz