Learn More
Andwin Scientific MICRO COVERGLASS 22X40 1 OZ
SDP
Supplier:  Andwin Scientific 7220030
Encompass_Preferred
Micro Cover Glasses Rectangular No. 1 No. 1 thickness is 0.13 to 0.17mm. 22x40mm. These cover glasses offer maximum clarity superior corrosion resistance and uniform thickness and size. They are free of stri and oily film., 1/OZ
Catalog No. 50-948-950