Learn More
Andwin Scientific MICRO COVERGLASS 22X50 1 OZ
SDP
Catalog No. 50948955
Encompass_Preferred
The micro cover glasses are made from clear white, borosilicate glass that provide superior clarity. This product has a size of 22 x 50 mm and thickness of 0.16-0.19 mm. It also features resistance against corrosion and a perfectly flat design. This is sold in a 1 ounce pack of 61 cover glasses.
Catalog No. 50-948-955 Supplier Andwin Scientific Supplier No. 7220404
The micro cover glasses are made from clear white, borosilicate glass that provide superior clarity. This product has a size of 22 x 50 mm and thickness of 0.16-0.19 mm. It also features resistance against corrosion and a perfectly flat design. This is sold in a 1 ounce pack of 61 cover glasses.