Learn More
Azzota Corp Micro Cuvette 1.4ml 40mm blk
Catalog No. 5000887132
Encompass_Preferred
Micro Cuvette 1 4ml 40mm Pathlength 1mm Inside Width Dimensions - 45x12 5x42 5mm Black walls Cover/Lid Included Quartz (190nm-2500nm)
Catalog No. 50-008-87132 Supplier Azzota Corp Supplier No. Q127
Micro Cuvette 1 4ml 40mm Pathlength 1mm Inside Width Dimensions - 45x12 5x42 5mm Black walls Cover/Lid Included Quartz (190nm-2500nm)