Learn More
Azzota Corp Micro Cuvette 2.1ml 20mm blk
Catalog No. 5000887032
Encompass_Preferred
MICRO CUVETTE 2 1ML 20MM PATHLENGTH 3MM INSIDE WIDTH Black Walls Quartz
Catalog No. 50-008-87032 Supplier Azzota Corp Supplier No. Q135
MICRO CUVETTE 2 1ML 20MM PATHLENGTH 3MM INSIDE WIDTH Black Walls Quartz