Learn More
Andwin Scientific Disposable Cuvette, 10L x 4W x 45mm H, 500/CS
SDP
Catalog No. NC9330837
Encompass_Preferred
Disposable Cuvette, Two optical windows semi-micro 1.5mL with 1.2mL minimum filling Volume and 10mm path Length. UV grade methyl acrylate for excellent light transmittance down to 275nm in the UV region. 10L x 4W x 45mm H, 500/CS
Catalog No. NC9330837 Supplier Andwin Scientific Supplier No. 58017850X
Disposable Cuvette, Two optical windows semi-micro 1.5mL with 1.2mL minimum filling Volume and 10mm path Length. UV grade methyl acrylate for excellent light transmittance down to 275nm in the UV region. 10L x 4W x 45mm H, 500/CS