Learn More
Starna Cells Inc MICRO CUVETTE / STOPPER .7ML
SDP
Supplier:  Starna Cells Inc 28Q10
Encompass_Preferred
Starna Micro Cells Stopper Top, Window material Spectrosil Quartz or equivalent, Path Length: 10mm, Ext Width: 12.5mm, Ext Length: 12.5mm, Ext Height: 48mm, Int Width: 2mm, Int Length: 10mm, Nominal Volume: 0.700mL
Catalog No. NC9028834