Learn More
Andwin Scientific MICRO-DILUTION TUBES PP STRI
SDP
Catalog No. 50819611
Encompass_Preferred
The 1.1 ml tubes in strips of 8 are compatible as refill racks for products products 1212-8400, 1212-8410, 1212-1600, or 1212-1610. There are 120 strips in this product, totaling to 960 tubes. All polypropylene tubes are in natural color.
This item is not returnable. View return policy
Catalog No. 50-819-611 Supplier Andwin Scientific Supplier No. 12128000
The 1.1 ml tubes in strips of 8 are compatible as refill racks for products products 1212-8400, 1212-8410, 1212-1600, or 1212-1610. There are 120 strips in this product, totaling to 960 tubes. All polypropylene tubes are in natural color.