Learn More
Roboz Surgical Extra Fine Micro Dissecting Scissors; 24 mm Blade
SDP
Catalog No. NC9232515
Encompass
Micro Dissecting Scissors; Straight; Sharp Points; 24mm Blade Length; 4 in. Overall Length
Catalog No. NC9232515 Supplier Roboz Surgical Supplier No. RS5882
Micro Dissecting Scissors; Straight; Sharp Points; 24mm Blade Length; 4 in. Overall Length