Learn More
Roboz Surgical Roboz Micro Dissecting Scissors, Curved, Sharp, 4.5 in, 1 ea
SDP
Catalog No. NC0212185
Encompass
These micro dissecting scissors are precision instruments featuring curved blades with sharp points, designed for delicate tissue cutting. The unique design incorporates one micro-serrated blade and one bevel-honed razor-sharp blade to ensure clean, effortless cuts without tissue crushing.
  • Features a 24mm blade length and 4 1/2-inch overall length
  • Combines a micro-serrated blade with a razor-sharp blade to prevent tissue slippage
  • Reduces the amount of force required during cutting procedures
  • Includes matte grey rings and a black screw for identification
  • Intended for soft tissue cutting only; avoid use on bone, wire, or fur to prevent blade damage
Catalog No. NC0212185 Supplier Roboz Surgical Supplier No. RS5913SC
These micro dissecting scissors are precision instruments featuring curved blades with sharp points, designed for delicate tissue cutting. The unique design incorporates one micro-serrated blade and one bevel-honed razor-sharp blade to ensure clean, effortless cuts without tissue crushing.
  • Features a 24mm blade length and 4 1/2-inch overall length
  • Combines a micro-serrated blade with a razor-sharp blade to prevent tissue slippage
  • Reduces the amount of force required during cutting procedures
  • Includes matte grey rings and a black screw for identification
  • Intended for soft tissue cutting only; avoid use on bone, wire, or fur to prevent blade damage