Learn More
Roboz Surgical Slight Curve Micro Dissecting Forceps
SDP
Catalog No. NC1681925
Encompass
Micro Dissecting Forceps; Serrated, Slight Curve; Extra Delicate; 0.5mm Tip Width; 4" Length
Catalog No. NC1681925 Supplier Roboz Surgical Supplier No. RS5136
Micro Dissecting Forceps; Serrated, Slight Curve; Extra Delicate; 0.5mm Tip Width; 4" Length