Learn More
Roboz Surgical MICRO DISS FCPS 4IN 1X2 STR .8
SDP
Catalog No. NC9267027
Encompass
Micro Dissecting Forceps; 1X2 Teeth, Straight; 0.8mm Tip Width; 4 in. Length
Catalog No. NC9267027 Supplier Roboz Surgical Supplier No. RS5150
Micro Dissecting Forceps; 1X2 Teeth, Straight; 0.8mm Tip Width; 4 in. Length