Learn More
Roboz Surgical MICRO DISS FCPS 4IN SER STR .5
SDP
Catalog No. NC9273606
Encompass
Micro Dissecting Forceps; Serrated, Straight; Extra Delicate; 0.5mm Tip Width; 4 in. Length
Catalog No. NC9273606 Supplier Roboz Surgical Supplier No. RS5132
Micro Dissecting Forceps; Serrated, Straight; Extra Delicate; 0.5mm Tip Width; 4 in. Length