Learn More
Roboz Surgical MICRO DISS SCS 4.5IN ST S/S TC
SDP
Catalog No. NC9585990
Encompass
Micro Dissecting Scissors; Sure Cut; Carbide Blades; Straight; Sharp Points; 24mm Blade Length; 4-1/2 in. Overall Length
Catalog No. NC9585990 Supplier Roboz Surgical Supplier No. RS5914SC
Micro Dissecting Scissors; Sure Cut; Carbide Blades; Straight; Sharp Points; 24mm Blade Length; 4-1/2 in. Overall Length