Learn More
Roboz Surgical MICRO DISS SCS 4.5IN STR B/B
SDP
Catalog No. NC9199223
Encompass
Micro Dissecting Scissors; Straight; Blunt; Standard Version; 25mm Blade Length; 4.5 in. Overall Length
Catalog No. NC9199223 Supplier Roboz Surgical Supplier No. RS5982
Micro Dissecting Scissors; Straight; Blunt; Standard Version; 25mm Blade Length; 4.5 in. Overall Length