Learn More
Roboz Surgical MICRO DISS VCPS 4IN F-CRV .5
SDP
Catalog No. NC9224242
Encompass
Micro Dissecting Forceps; Serrated, Full Curve; Extra Delicate; 0.5mm Tip Width; 4 in. Length
Catalog No. NC9224242 Supplier Roboz Surgical Supplier No. RS5138
Micro Dissecting Forceps; Serrated, Full Curve; Extra Delicate; 0.5mm Tip Width; 4 in. Length