Learn More
Roboz Surgical MICRO DISSECT TWEEZER STRGHT
SDP
Catalog No. NC9832366
Encompass
Dumont Tweezers; Pattern #5; .05 X .01mm Tip Size; 110mm Length; Material Inox; Biologie Tips
Catalog No. NC9832366 Supplier Roboz Surgical Supplier No. RS5015
Dumont Tweezers; Pattern #5; .05 X .01mm Tip Size; 110mm Length; Material Inox; Biologie Tips