Learn More
Roboz Surgical Roboz Surgical Micro Dissecting Scissors, 4.5 in, Ceramic, 1 ea
SDP
Catalog No. NC1953506
Encompass
These 4.5-inch micro dissecting scissors are designed for precision laboratory and surgical tasks, offering enhanced cutting performance and durability. Featuring tungsten carbide blades with a ceramic coating, these straight, sharp/sharp scissors incorporate a serrated design to securely grip delicate tissue and prevent slippage during use.
  • Tungsten carbide construction provides exceptional edge retention
  • Ceramic coating increases surface hardness for superior wear resistance
  • Serrated blade design ensures stable, controlled tissue engagement
  • Maintains sharp cutting edges over six times longer than standard scissors
Catalog No. NC1953506 Supplier Roboz Surgical Supplier No. RS5916
These 4.5-inch micro dissecting scissors are designed for precision laboratory and surgical tasks, offering enhanced cutting performance and durability. Featuring tungsten carbide blades with a ceramic coating, these straight, sharp/sharp scissors incorporate a serrated design to securely grip delicate tissue and prevent slippage during use.
  • Tungsten carbide construction provides exceptional edge retention
  • Ceramic coating increases surface hardness for superior wear resistance
  • Serrated blade design ensures stable, controlled tissue engagement
  • Maintains sharp cutting edges over six times longer than standard scissors