Learn More
Roboz Surgical MICRO DISSECTING FORCEP ANGLED
SDP
Catalog No. NC9791959
Encompass
Micro Dissecting Forceps; Angled; 11mm Tip Length; Smooth Jaws; 6 in. Length
Catalog No. NC9791959 Supplier Roboz Surgical Supplier No. RS5359
Micro Dissecting Forceps; Angled; 11mm Tip Length; Smooth Jaws; 6 in. Length