Learn More
Electron Microscopy Sciences Micro Dissecting Forceps Smooth Jaws with Curved Blades
SDP
Catalog No. 5034617
Encompass_Preferred
20 gauge stainless steel, with 3 mm tips.
Blades: Curved
Catalog No. 50-346-17 Supplier Electron Microscopy Sciences Supplier No. 7793203
20 gauge stainless steel, with 3 mm tips.
Blades: Curved