Learn More
Roboz Surgical MICRO DISSECTING SCISSOR
SDP
Catalog No. NC9178691
Encompass
Castroviejo Micro Dissecting Spring Scissors; Curved, Sharp Points; 10mm Cutting Edge; 0.15mm Tip Width; 3-3/4 in. Overall Length
Catalog No. NC9178691 Supplier Roboz Surgical Supplier No. RS5671
Castroviejo Micro Dissecting Spring Scissors; Curved, Sharp Points; 10mm Cutting Edge; 0.15mm Tip Width; 3-3/4 in. Overall Length