Learn More
Roboz Surgical MICRO DISSECTING SCISSOR
SDP
Catalog No. NC9178689
Encompass
Vannas Micro Dissecting Spring Scissors; Curved; 3mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length These scissors have finer blades than the very similar RS-5611 scissors. The tips are narrower and the characteristic notch in the blades of this
Catalog No. NC9178689 Supplier Roboz Surgical Supplier No. RS5621
Vannas Micro Dissecting Spring Scissors; Curved; 3mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length These scissors have finer blades than the very similar RS-5611 scissors. The tips are narrower and the characteristic notch in the blades of this