Learn More
Roboz Surgical Roboz Surgical SureCut Dissecting Scissors, Straight, 3.5 in, 1 ea
SDP
Catalog No. NC1211655
Encompass
These micro dissecting scissors are precision instruments designed for delicate tissue handling and fine surgical procedures. Featuring high-durability tungsten carbide inserts and advanced cutting technology, they ensure long-lasting sharpness and reliable performance in laboratory or clinical environments.
  • Features tungsten carbide cutting edges for extended service life
  • Designed with a 3.5-inch total length and sharp, straight tips
  • Utilizes specialized technology to maintain a clean, precise cut
  • Includes a warning to avoid cutting wire, thread, or other hard materials to protect the blade integrity
  • Equipped with a distinct black screw and matte grey ring finish
This item is not returnable. View return policy
Catalog No. NC1211655 Supplier Roboz Surgical Supplier No. RS5906SC
These micro dissecting scissors are precision instruments designed for delicate tissue handling and fine surgical procedures. Featuring high-durability tungsten carbide inserts and advanced cutting technology, they ensure long-lasting sharpness and reliable performance in laboratory or clinical environments.
  • Features tungsten carbide cutting edges for extended service life
  • Designed with a 3.5-inch total length and sharp, straight tips
  • Utilizes specialized technology to maintain a clean, precise cut
  • Includes a warning to avoid cutting wire, thread, or other hard materials to protect the blade integrity
  • Equipped with a distinct black screw and matte grey ring finish