Learn More
Roboz Surgical MICRO DISSECTING SCISSORS
SDP
Catalog No. NC1509648
Encompass
These micro dissecting scissors are designed for delicate surgical procedures, featuring a 3.5 in. length and a curved design with sharp/sharp tips. Equipped with tungsten carbide inserts for enhanced durability and precision, these scissors are suitable for fine-scale tissue manipulation and micro-dissection tasks.
  • Tungsten carbide inserts provide long-lasting sharpness and wear resistance
  • Curved geometry allows for improved visibility and access in confined spaces
  • Sharp/sharp tips enable precise point-to-point dissection
  • Gold ring handles assist in easy identification
  • Overall length of 3.5 in. offers superior control for intricate surgical work
Catalog No. NC1509648 Supplier Roboz Surgical Supplier No. RS5908
These micro dissecting scissors are designed for delicate surgical procedures, featuring a 3.5 in. length and a curved design with sharp/sharp tips. Equipped with tungsten carbide inserts for enhanced durability and precision, these scissors are suitable for fine-scale tissue manipulation and micro-dissection tasks.
  • Tungsten carbide inserts provide long-lasting sharpness and wear resistance
  • Curved geometry allows for improved visibility and access in confined spaces
  • Sharp/sharp tips enable precise point-to-point dissection
  • Gold ring handles assist in easy identification
  • Overall length of 3.5 in. offers superior control for intricate surgical work