Learn More
Roboz Surgical Roboz SureCut Micro Scissors, 3.5 Inch, Straight, 1 ea
SDP
Catalog No. NC0648483
Encompass
These 3.5-inch straight micro dissecting scissors feature sharp/sharp tips and are engineered for high-precision surgical procedures. Constructed with tungsten carbide inserts and a specialized ceramic coating, these instruments offer enhanced durability, hardness, and a non-stick, glare-free surface to facilitate precise tissue manipulation.
  • Proprietary manufacturing process ensures longer-lasting sharpness and precise cutting performance.
  • Tungsten carbide inserts provide superior durability compared to standard stainless steel.
  • Ceramic coating creates a non-corrosive, non-stick, and bio-compatible surface.
  • Fine, sharp tips are carefully aligned to minimize tissue slippage during use.
  • Super-honed blades deliver clean and accurate cuts.
  • High surface hardness improves scratch resistance and longevity.
  • Non-stick finish allows for easy cleaning and maintenance.
Catalog No. NC0648483 Supplier Roboz Surgical Supplier No. RS5907
These 3.5-inch straight micro dissecting scissors feature sharp/sharp tips and are engineered for high-precision surgical procedures. Constructed with tungsten carbide inserts and a specialized ceramic coating, these instruments offer enhanced durability, hardness, and a non-stick, glare-free surface to facilitate precise tissue manipulation.
  • Proprietary manufacturing process ensures longer-lasting sharpness and precise cutting performance.
  • Tungsten carbide inserts provide superior durability compared to standard stainless steel.
  • Ceramic coating creates a non-corrosive, non-stick, and bio-compatible surface.
  • Fine, sharp tips are carefully aligned to minimize tissue slippage during use.
  • Super-honed blades deliver clean and accurate cuts.
  • High surface hardness improves scratch resistance and longevity.
  • Non-stick finish allows for easy cleaning and maintenance.