Learn More
Roboz Surgical Micro Dissecting Scissors
SDP
Catalog No. NC1674341
Encompass
Micro Dissecting Scissors; Straight; Sharp Points; 23mm Blade Length; Combined Tip Width: 0.8mm; 3 1/2" Overall Length
This item is not returnable. View return policy
Catalog No. NC1674341 Supplier Roboz Surgical Supplier No. RS5910
Micro Dissecting Scissors; Straight; Sharp Points; 23mm Blade Length; Combined Tip Width: 0.8mm; 3 1/2" Overall Length