Learn More
Roboz Surgical MICRO-DISSECTING SCISSORS
SDP
Catalog No. NC9337891
Encompass
Micro Dissecting Scissors; Carbide Blades; Straight; Sharp Points; 24mm Blade Length; 4-1/2 in. Overall Length
Catalog No. NC9337891 Supplier Roboz Surgical Supplier No. RS5914
Micro Dissecting Scissors; Carbide Blades; Straight; Sharp Points; 24mm Blade Length; 4-1/2 in. Overall Length