Learn More
Roboz Surgical MICRO DISSECTING SCISSORS
SDP
Catalog No. NC9581049
Encompass
These micro dissecting scissors are precision instruments designed for fine surgical procedures and delicate tissue manipulation. Featuring a curved design with sharp points, they allow for accurate access and cutting in confined spaces.
  • Curved geometry facilitates access in restricted areas
  • Sharp, precise points ensure clean incisions
  • Overall length of 3 1/2 in. for controlled handling
  • Blade length of 23 mm optimized for micro-dissection tasks
Catalog No. NC9581049 Supplier Roboz Surgical Supplier No. RS5911
These micro dissecting scissors are precision instruments designed for fine surgical procedures and delicate tissue manipulation. Featuring a curved design with sharp points, they allow for accurate access and cutting in confined spaces.
  • Curved geometry facilitates access in restricted areas
  • Sharp, precise points ensure clean incisions
  • Overall length of 3 1/2 in. for controlled handling
  • Blade length of 23 mm optimized for micro-dissection tasks