Learn More
Roboz Surgical MICRO DISSECTING TWEEZERS (TIP
SDP
Catalog No. NC9208902
Encompass
Dumont Tweezers; Pattern #7; .17 X .10mm Tip Size; 115mm Length; Material: Dumostar
Catalog No. NC9208902 Supplier Roboz Surgical Supplier No. RS4980
Dumont Tweezers; Pattern #7; .17 X .10mm Tip Size; 115mm Length; Material: Dumostar