Learn More
Roboz Surgical MICRO DISSECTION TWEEZERS
SDP
Catalog No. NC9535535
Encompass
Dumont Tweezers; Pattern #2; .34 X .14mm Tip Size; 120mm Length; Material: Dumostar
Catalog No. NC9535535 Supplier Roboz Surgical Supplier No. RS4962
Dumont Tweezers; Pattern #2; .34 X .14mm Tip Size; 120mm Length; Material: Dumostar